missing translation for 'onlineSavingsMsg'
Learn More

AL 8697, MedChemExpress

Product Code. 16476937
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476937 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16476937 Supplier Medchem Express Supplier No. HY108645
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AL 8697 is a specific and orally active p38α MAPK inhibitor with an IC50 of 6 nM. AL 8697 displays 14-fold greater inhibition of p38α compared to p38β (IC50=82 nM), and 300-fold selectivity for p38α over a panel of 91 kinases. Anti-inflammatory activity.

AL 8697 is a specific and orally active p38α MAPK inhibitor with an IC50 of 6 nM. AL 8697 displays 14-fold greater inhibition of p38α compared to p38β (IC50=82 nM), and 300-fold selectivity for p38α over a panel of 91 kinases. Anti-inflammatory activity.

Specifications

Chemical Name or Material AL 8697
CAS 1057394-06-5
Color White
Purity Grade Notes Research
Molecular Formula C21H21F3N4O
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (248.50 mM; Need ultrasonic)
SMILES O=C(NC1CC1)C2=CC(F)=C(C)C(C3=C(F)C4=NN=C(C(C)(C)C)N4C=C3F)=C2
Molecular Weight (g/mol) 402.41
Formula Weight 402.41
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.