missing translation for 'onlineSavingsMsg'
Learn More

Al-24 Tube, Both ends open, O.D.: 12mm, I.D.: 8mm, Alfa Aesar™

Brand:  Alfa Aesar 031602.DW

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300mm

 View more versions of this product

Product Code. 11324087

  • £46.50 / 300mm


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Al-24 Tube
Both ends open

Specifications

Al-24 Tube
300mm
Both ends open
Videos
SDS