missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Al-23 Tube, One End Closed, O.D.: 55mm, I.D.: 47mm, Alfa Aesar™

Brand:  Thermo Scientific Alfa Aesar 030085.DM

Al-23 Tube, One End Closed, O.D.: 55mm, I.D.: 47mm, Alfa Aesar™

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11332838

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.