missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, One End Closed, O.D.: 4mm, I.D.: 2mm, Alfa Aesar™

Brand:  Alfa Aesar 030062.DR

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 200mm

 View more versions of this product

Product Code. 11362718

  • £49.30 / 200mm


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
One end closed

Specifications

Al-23 Tube
200mm
One end closed
Videos
SDS