missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Al-23 Tube, One End Closed, O.D.: 35mm, I.D.: 29mm, Alfa Aesar™

Brand:  Thermo Scientific Alfa Aesar 030081.DM

Al-23 Tube, One End Closed, O.D.: 35mm, I.D.: 29mm, Alfa Aesar™

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11362808

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers