missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both Ends Open, O.D.: 15mm, I.D.: 10mm, Alfa Aesar™

Brand:  Alfa Aesar 031566.EO

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1000mm

 View more versions of this product

Product Code. 11303977

  • £156.46 / 1000mm


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
Both ends open

Specifications

Al-23 Tube
1000mm
Both ends open
Videos
SDS