missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both Ends Open, O.D.: 10mm, I.D.: 6mm, Alfa Aesar™

Brand:  Alfa Aesar 031562.DM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100mm

 View more versions of this product

Product Code. 11333957

  • £27.10 / 100mm


Estimated Shipment date
from Supplier 25-01-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Tube
Both ends open

Specifications

Al-23 Tube
100mm
Both ends open
Videos
SDS