missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Solid Round Rod, Dia.: 0.8mm, Alfa Aesar™

Brand:  Alfa Aesar 032922.DM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100mm

 View more versions of this product

Product Code. 11353038

  • £16.20 / 100mm


Estimated Shipment date
from Supplier 25-01-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Solid Round Rod

Specifications

Al-23 Solid Round Rod
100mm
Videos
SDS