missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Solid Rod, Round, Dia.: 4mm, Alfa Aesar™

Brand:  Alfa Aesar 032929.DM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100mm

 View more versions of this product

Product Code. 11363068

  • £16.00 / 100mm


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Solid Rod
Solid Round Rod

Specifications

Al-23 Solid Rod
100mm
Solid Round Rod
Videos
SDS