missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Protective Tube, One End Closed, O.D.: 12mm, I.D.: 8mm, Wall.: 2mm, Thermo Scientific™

Product Code. 11372678
Change view
Click to view available options
Quantity:
200 mm
270 mm
375 mm
530 mm
740 mm
1030 mm
1460 mm
2030 mm
Unit Size:
1030mm
1460mm
200mm
2030mm
270mm
375mm
530mm
740mm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372678 375 mm 375mm
11342688 200 mm 200mm
11322688 270 mm 270mm
11302688 530 mm 530mm
11392678 740 mm 740mm
11332688 1030 mm 1030mm
15471137 1460 mm 1460mm
11382678 2030 mm 2030mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 11372678 Supplier Thermo Scientific Alfa Aesar Supplier No. 030048.DY
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Al-23 Protective Tube
Name Note One end closed
Quantity 375 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.