missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Oval Cross Section 2-Bore Insulators, O.D.: 4.2mm/2.2mm, Two 1.2mm Bores, Alfa Aesar7trade;

Brand:  Alfa Aesar 032775.DM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100mm

 View more versions of this product

Product Code. 11382998

  • £22.40 / 100mm


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Oval Cross Section 2-Bore Insulator
Two bores

Specifications

Al-23 Oval Cross Section 2-Bore Insulator
100mm
Two bores
Videos
SDS