missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Multibore Round Tube, O.D.: 4mm, Two 0.8mm Bores, Alfa Aesar™

Brand:  Alfa Aesar 032650.DM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100mm

 View more versions of this product

Product Code. 11352898

  • £11.40 / 100mm


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Multibore Round Tube
Two bores

Specifications

Al-23 Multibore Round Tube
100mm
Two bores
Videos
SDS