missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Multibore Round Tube, O.D.: 0.9mm, Two 0.2mm Bores, Alfa Aesar™

Brand:  Alfa Aesar 032564.DM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100mm

 View more versions of this product

Product Code. 11338467

  • £15.70 / 100mm


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

Al-23 Multibore Round Tube
Two bores

Specifications

Al-23 Multibore Round Tube
100mm
Two bores
Videos
SDS