missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Al-23 Multibore Insulating Tube, O. D.: 6mm, Two 1.5mm Bores, Thermo Scientific™
Click to view available options
Quantity:
205 mm
275 mm
380 mm
560 mm
770 mm
1060 mm
1460 mm
2060 mm
Unit Size:
1060mm
1460mm
205mm
2060mm
275mm
380mm
560mm
770mm
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Chemical Name or Material | Al-23 Multibore Insulating Tube |
| Name Note | Two bores |
| Quantity | 380 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction