missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Multibore Insulating Tube, O. D.: 6mm, Two 1.5mm Bores, Thermo Scientific™

Product Code. 11388437
Change view
Click to view available options
Quantity:
205 mm
275 mm
380 mm
560 mm
770 mm
1060 mm
1460 mm
2060 mm
Unit Size:
1060mm
1460mm
205mm
2060mm
275mm
380mm
560mm
770mm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388437 2060 mm 2060mm
11358437 205 mm 205mm
11378437 275 mm 275mm
11318447 380 mm 380mm
11308447 560 mm 560mm
11348437 770 mm 770mm
11368437 1060 mm 1060mm
11398437 1460 mm 1460mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 11388437 Supplier Thermo Scientific Alfa Aesar Supplier No. 032556.FB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Al-23 Multibore Insulating Tube
Name Note Two bores
Quantity 2060 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.