missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Multibore Insulating Tube, O. D.: 4mm, Four 0.8mm Bores, Thermo Scientific™

Product Code. 11328427
Change view
Click to view available options
Quantity:
205 mm
275 mm
380 mm
560 mm
770 mm
1060 mm
1460 mm
2060 mm
Förpackningsstorlek:
1060mm
1460mm
205mm
2060mm
275mm
380mm
560mm
770mm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
11328427 205 mm 205mm
11398417 275 mm 275mm
11388417 380 mm 380mm
11308427 560 mm 560mm
11358427 770 mm 770mm
11318427 1060 mm 1060mm
11348427 1460 mm 1460mm
11338427 2060 mm 2060mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11328427 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer 032552.DS
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Al-23 Multibore Insulating Tube
Name Note Four bores
Quantity 205 mm
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.