missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Insulating Tube, Both Ends Open (Thin Wall), O.D.: 0.5mm, I.D.: 0.2mm, Thermo Scientific Chemicals

POA - £35.20

Specifications

Chemical Name or Material Al-23 Insulating Tube
Name Note Both ends open (Thin Wall)
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11397468
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031475.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11327478
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031475.DR
200 mm
£20.20
200mm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11317478
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031475.DT
250 mm
£26.90
250mm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11307478
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031475.DW
300 mm
£35.20
300mm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Al-23 Insulating Tube
Both ends open (Thin Wall)
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : U

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only