missing translation for 'onlineSavingsMsg'
Learn More

AL 082D06, MedChemExpress

Product Code. 16489307
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489307 5 mg 5mg
16476886 100 mg 100mg
16476758 10 mg 10mg
16472837 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489307 Supplier Medchem Express Supplier No. HY15709
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AL 082D06 is a selective, nonsteroidal glucocorticoid receptor (GR) antagonist with Ki of 210 nM.

AL 082D06 is a selective, nonsteroidal glucocorticoid receptor (GR) antagonist with Ki of 210 nM.

Specifications

Chemical Name or Material AL 082D06
Physical Form Solid
CAS 256925-03-8
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H24ClN3O2
Synonym D06 D-06
Solubility Information DMSO : 7.5 mg/mL (18.30 mM; Need ultrasonic and warming)
SMILES O=[N+](C1=CC=C(Cl)C(C(C2=CC=C(N(C)C)C=C2)C3=CC=C(N(C)C)C=C3)=C1)[O-]
Molecular Weight (g/mol) 409.91
Formula Weight 409.91
Percent Purity 98.92%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.