missing translation for 'onlineSavingsMsg'
Learn More

Cytiva ÄKTA™ High Flow Kit

Higher flow rates at lower back pressure. Cytiva ÄKTA™ High Flow Kit contains the necessary components and software to reduce backpressure in the system, thereby allowing higher flow rates.

Brand:  Cytiva 18-1154-24

Product Code. 11380262

  • £929.00 / Each
Estimated Shipment: 04-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The kit contains the necessary components and software to reduce backpressure in the system, and thereby allowing higher flow rates. This makes it a suitable setup for running larger columns and for increasing the production capacity.
  • Higher flow rate: 200mL/min
  • Lower back pressure: <0.6 MPa
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Increasing Flow Rate While Lowering Back Pressure
<0.6 MPa
Includes connectors, tubing, ferules and basic tools
ÄKTAexplorer 100 System, Esp. with Larger Columns
Max. flow rate: 200 mL/min.
1 Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers