missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT3 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21641-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the AKT, also called PKB, serine/threonine protein kinase family. AKT kinases are known to be regulators of cell signaling in response to insulin and growth factors. They are involved in a wide variety of biological processes including cell proliferation, differentiation, apoptosis, tumorigenesis, as well as glycogen synthesis and glucose uptake. This kinase has been shown to be stimulated by platelet-derived growth factor (PDGF), insulin, and insulin-like growth factor 1 (IGF1). Alternatively splice transcript variants encoding distinct isoforms have been described.Specifications
| AKT3 | |
| Polyclonal | |
| Unconjugated | |
| AKT3 | |
| AKT3, PKB GAMMA, PKBG, PRKBG, Protein kinase Akt 3, Protein kinase B gamma, RAC gamma, RAC PK gamma, STK 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10000, 23797, 29414, 560549 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q63484, Q9WUA6, Q9Y243 | |
| Akt3, akt3a | |
| AKT3 Fusion Protein Ag16298 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction