missing translation for 'onlineSavingsMsg'
Learn More

AKT3 Antibody (3F3), Novus Biologicals™

Product Code. 18328499 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328499 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328499 Supplier Novus Biologicals Supplier No. H00010000M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AKT3 Monoclonal antibody specifically detects AKT3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKT3
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3F3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAD29089
Gene Alias Akt-3, EC 2.7.11, EC 2.7.11.1, PKB gamma, PKBGDKFZp434N0250, PRKBG, v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma)
Host Species Mouse
Immunogen AKT3 (AAD29089, 100 a.a. ~ 189 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EAIQAVADRLQRQEEERMNCSPTSQIDNIGEEEMDASTTHHKRKTMNDFDYLKLLGKGTFGKVILVREKASGKYYAMKILKKEVIIAKDE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10000
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.