missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT3 Antibody (199822) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB14631P
This item is not returnable.
View return policy
Description
AKT3 Monoclonal antibody specifically detects AKT3 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| AKT3 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 199822 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Akt-3, EC 2.7.11, EC 2.7.11.1, PKB gamma, PKBGDKFZp434N0250, PRKBG, v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma) | |
| E. coli-derived recombinant human Akt3, Arg23-Ala101, Accession # NP_005456 | |
| 0.1 mL | |
| Cancer, Hypoxia, Protein Kinase, Signal Transduction | |
| 10000 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction