Learn More
Invitrogen™ AKT2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA595178
Product Code. 16374445
  • £441.00 / 100µg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human SW620 whole cell, human 293T whole cell, human MCF-7 whole cell, rat brain tissue, mouse brain tissue. IHC: human lung cancer tissue, human intestinal cancer tissue IHC-F: rat brain tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
AKT2 is a phosphoinositide-dependent serine/threonine protein kinase that is enriched in insulin responsive tissues and has been implicated in the metabolic actions of the hormone. Akt is involved in many cellular signaling pathways and acts as a transducer of many functions initiated by growth factor receptors that activate phosphatidylinositol 3-kinase (PI 3-kinase). Akt2 is amplified and overexpressed in some human cancers.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AKT2
Polyclonal
Unconjugated
Akt2
2410016A19Rik; AKT serine/threonine kinase 2; Akt2; AKT2 kinase; AW554154; HIHGHH; murine thymoma viral (v-akt) homolog-2; murine thymoma viral (v-akt) oncogene homolog 2; PKB; PKB beta; PKBB; PKBBETA; PRKBB; protein kinase Akt-2; Protein kinase B beta; protein kinase B, beta; putative v-akt murine thymoma viral oncoprotein 2; rac protein kinase beta; RAC protein kinase beta RAC-PK beta; RAC-BETA; RAC-beta serine/threonine protein kinase; RAC-beta serine/threonine-protein kinase; RAC-PK-beta; thymoma viral proto-oncogene 2; v-akt murine thymoma viral oncogene homolog 2
Rabbit
Affinity chromatography
RUO
11652, 208, 25233
-20°C
Lyophilized
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
500 μg/mL
PBS with 5mg BSA and 0.05mg sodium azide
P31751, P47197, Q60823
Akt2
A synthetic peptide corresponding to a sequence at the C-terminus of human AKT2 (454-481aa DRYDSLGLLELDQRTHFPQFSYSASIRE).
100 μg
Primary
Human, Mouse, Rat