Learn More
Invitrogen™ Human AKT2 (PKB beta), His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
Specifications
Specifications
| Accession Number | P31751 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 20 mM tris with 0.01% Triton X-100, 0.1 mM sodium orthovanadate, 0.5 mM EDTA, 150 mM NaCl, 20% glycerol, 2 mM DTT and no preservative; pH 7.5 |
| Gene ID (Entrez) | 208 |
| Molecular Weight (g/mol) | 59.1 kDa |
| Name | Human AKT2 (PKB beta), His Tag |
| Quantity | 100 μg |
| Regulatory Status | RUO - research use only |
| Gene Alias | 2410016A19Rik; AKT serine/threonine kinase 2; Akt2; AKT2 kinase; AW554154; HIHGHH; murine thymoma viral (v-akt) homolog-2; murine thymoma viral (v-akt) oncogene homolog 2; PKB; PKB beta; PKBB; PKBBETA; PRKBB; protein kinase Akt-2; Protein kinase B beta; protein kinase B, beta; putative v-akt murine thymoma viral oncoprotein 2; rac protein kinase beta; RAC protein kinase beta RAC-Pk. beta; RAC-BETA; RAC-beta serine/threonine protein kinase; RAC-beta serine/threonine-protein kinase; RAC-Pk.-beta; thymoma viral proto-oncogene 2; v-akt murine thymoma viral oncogene homolog 2 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.