missing translation for 'onlineSavingsMsg'
Learn More

AKT2 Antibody (1D9), Novus Biologicals™

Product Code. 18396537 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396537 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396537 Supplier Novus Biologicals Supplier No. H00000208M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AKT2 Monoclonal antibody specifically detects AKT2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKT2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1D9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAA58364
Gene Alias EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2
Host Species Mouse
Immunogen AKT2 (AAA58364, 100 a.a. ~ 189 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MRAIQMVANSLKQRAPGEDPMDYKCGSPSDSSTTEEMEVAVSKARAKVTMNDFDYLKLLGKGTFGKVILVREKATGRYYAMKILRKEVII
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Hypoxia, mTOR Pathway, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 208
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.