missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT2 Antibody (11F6.B2), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-44216
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, ELISA, Flow Cytometry, Immunohistochemistry
AKT2 Monoclonal specifically detects AKT2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.
Specifications
| AKT2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:2000-1:10000, Immunohistochemistry 20 ug/ml | |
| P31751, P31751, P31751, P31751 | |
| AKT2 | |
| AKT2 Antibody (11F6.B2) was produced by repeated immunizations with a synthetic peptide corresponding to internal residues of human AKT2 protein. (Uniprot: P31751) | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-AKT2 antibody is directed against human AKT2. The antibody detects both unphosphorylated and phosphorylated forms of the protein. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 11F6.B2 | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2 | |
| Rat | |
| 55.77 kDa | |
| 0.1 mg | |
| Apoptosis, Cancer, Hypoxia, mTOR Pathway, Protein Kinase, Signal Transduction | |
| 208 | |
| Human, C. elegans | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction