missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1-Specific Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80816-1-RR-20UL
This item is not returnable.
View return policy
Description
The AKT1-Specific antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human AKT1-Specific. This antibody recognizes human, mouse, rat antigen. The AKT1-Specific antibody has been validated for the following applications: WB, IHC, FC (Intra), IP, ELISA analysis.Specifications
| AKT1-Specific | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| AKT; MGC99656; PKB; PKB-ALPHA; PRKBA; RAC; RAC-ALPHA | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 56-62 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunoprecipitation, ELISA | |
| 5O9 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 207 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction