missing translation for 'onlineSavingsMsg'
Learn More

Akt1 and Akt2-IN-1, MedChemExpress

Product Code. 16487876
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487876 100 mg 100mg
16450578 5 mg 5mg
16448118 10 mg 10mg
16424037 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16487876 Supplier Medchem Express Supplier No. HY50862
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Akt1 and Akt2-IN-1 is an allosteric inhibitor of Akt1 (IC50=3.5 nM) and Akt2 (IC50=42 nM), with potent and balanced activity.

Akt1 and Akt2-IN-1 is an allosteric inhibitor of Akt1 (IC50=3.5 nM) and Akt2 (IC50=42 nM), with potent and balanced activity.

Specifications

Chemical Name or Material Akt1 and Akt2-IN-1
CAS 893422-47-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C33H29N7O
Quantity 100 mg
Solubility Information DMSO : ≥ 35 mg/mL (64.86 mM)
SMILES O=C(NC=C1)C2=C1N=C(C(C3=CC=CC=C3)=C2)C(C=C4)=CC=C4CN5CCC(CC5)C6=NNC(C7=NC=CC=C7)=N6
Molecular Weight (g/mol) 539.63
Formula Weight 539.63
Percent Purity 99.59%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.