missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1/2/3 Rabbit anti-Human, Mouse, Rat, Clone: 2E3A3, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £423.00
Specifications
| Antigen | AKT1/2/3 |
|---|---|
| Clone | 2E3A3 |
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18386854
|
Novus Biologicals
NBP3-16948-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18342257
|
Novus Biologicals
NBP3-16948-100UL |
100 μg |
£423.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AKT1/2/3 Monoclonal antibody specifically detects AKT1/2/3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| AKT1/2/3 | |
| Western Blot 1:500 - 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AKT, EC 2.7.11, EC 2.7.11.1, PKBMGC99656, PRKBA, Protein kinase B, Proto-oncogene c-Akt, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinase, RAC-PK-alpha, RACPKB-ALPHA, v-akt murine thymoma viral oncogene homolog 1 | |
| A synthetic phosphorylated peptide around S473 of human AKT1/2/3 (NP_005154.2). QFSYS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 2E3A3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 207 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title