missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT [p Ser473] Antibody (545007) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB887N
427.55 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
AKT Monoclonal antibody specifically detects AKT in Human, Mouse samples. It is validated for Western Blot, Simple Western
Specifications
| AKT | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Simple Western | |
| 545007 | |
| Western Blot, Simple Western | |
| RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC | |
| Phosphopeptide containing the human Akt (S473) site | |
| 0.1 mL | |
| Cellular Signaling, Neuroscience | |
| 207 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction