missing translation for 'onlineSavingsMsg'
Learn More

AKT [p Ser473] Antibody (545007) [Alexa Fluor™ 594], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB887T

427.55 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18559478

  • £536.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AKT Monoclonal antibody specifically detects AKT in Human, Mouse samples. It is validated for Western Blot, Simple Western
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AKT
Monoclonal
Alexa Fluor 594
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human, Mouse
Purified
Western Blot, Simple Western
545007
Western Blot, Simple Western
RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC
Phosphopeptide containing the human Akt (S473) site
0.1 mL
Cellular Signaling, Neuroscience
207
Store at 4°C in the dark.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.