missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT Mouse anti-Human, Mouse, Rat, Clone: 2C5D1, Proteintech
Mouse Monoclonal Antibody
£130.00 - £349.00
Specifications
| Antigen | AKT |
|---|---|
| Clone | 2C5D1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16847205
|
Proteintech
60203-2-IG-20UL |
20 μL |
£135.00 £130.00 / 20µL Save £5.00 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16837205
|
Proteintech
60203-2-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody detects all the members of AKT with/without phospho-modification.
Akt1 gene ontology annotations related to this gene include intracellular signal transduction.Specifications
| AKT | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| AKT, AKT (pan), AKT pan, AKT1, AKT1/2/3, pan AKT, PKB, PKB ALPHA, PRKBA, Protein kinase B, Proto oncogene c Akt, RAC, RAC ALPHA, RAC PK alpha, total AKT | |
| AKT1, AKT2, Akt3 | |
| IgG1 | |
| Protein A | |
| Antibody |
| 2C5D1 | |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Rat, Mouse, Human | |
| P31749, P31750, P31751, P47196, P47197, Q60823, Q63484, Q9WUA6, Q9Y243 | |
| 10000, 11651, 11652, 207, 208, 23797, 24185, 25233, 29414 | |
| AKT Fusion Protein Ag16695 | |
| Primary | |
| -20°C | |
| AKT1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title