missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEA0400, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
Description
SEA0400 is a novel and selective inhibitor of the Na+-Ca2+ exchanger (NCX), inhibiting Na+-dependent Ca2+ uptake in cultured neurons, astrocytes, and microglia with IC50s of from 5 to 33 nM.
Specifications
Specifications
| Chemical Name or Material | SEA0400 |
| CAS | 223104-29-8 |
| Color | Khaki |
| Purity Grade Notes | Research |
| Molecular Formula | C21H19F2NO3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 32 mg/mL (86.17 mM) |
| SMILES | NC1=CC(OCC)=CC=C1OC2=CC=C(OCC3=CC(F)=CC=C3F)C=C2 |
| Molecular Weight (g/mol) | 371.38 |
| Formula Weight | 371.38 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction