missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies AKT Inhibitor VIII

Product Code. 17110152
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 17110152

Brand: STEMCELL Technologies 72942

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PI3K/AKT pathway inhibitor; Inhibits AKT1, AKT2, and AKT3

  • AKT Inhibitor VIII is a cell-permeable, allosteric inhibitor of all three forms of the kinase AKT (AKT1, AKT2, and AKT3) with IC50 values of 58, 210, and 2,200 nM, respectively (Lindsley et al.; Calleja et al.) It displays good selectivity against a panel of 70 other kinases with micromolar inhibition against some kinases, such as calcium/calmodulin-dependent protein kinase 1 and smooth muscle myosin light-chain kinase (Logie et al.)
  • Sensitizes prostate tumor and cervical carcinoma cells to apoptotic stimuli (DeFeo-Jones et al.)
  • Blocks mitosis and inhibits migration of HeLa cells (Jo et al.)
  • Reduces insulin-dependent gene repression in liver cells leading to reduced insulin sensitivity (Logie et al.)

Specifications

CAS 612847-09-3
Synonym AKTi-1/2; AKT 1/2 inhibitor
Quantity 1 mg
Molecular Formula C34H29N7O
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.