missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AKT Inhibitor IV

Description
AKT Inhibitor IV promotes hyperphosphorylation of Akt (Protein kinase B), decreases phosphorylation of Akt substrates, and inhibits FOXO1a nuclear export (IC50 = 625 nM in PTEN-null cells). AKT Inhibitor IV inhibits the proliferation of several cells lines (IC50 values are <1.25 μM, 320 - 670 nM and 340 nM for 786-O, HeLa and Jurkat cells, respectively). This compound also displays broad antiviral activity against negative-stranded RNA viruses; inhibits replication of parainfluenza virus 5 (PIV5) in HeLa cells (IC50 = 520 nM). In HeLa cells, AKT Inhibitor IV accumulates in mitochondria, disrupting morphology and increasing ROS production. At high concentrations (10 μM), AKT Inhibitor IV activates the unfolded protein response and apoptosis in HEK293T cells.
AKT Inhibitor IV is weakly fluorescent upon irradiation with UV light (excitation/emission maxima λ ∼ 388/460; quantum yield = 0.0016).
Specifications
Specifications
| Chemical Name or Material | 6-(2-Benzothiazolyl)-1-ethyl-2-[(1 E)-2-(methylphenylamino)ethenyl]-3-phenyl-1 H-benzimidazolium iodide |
| CAS | 959841-49-7 |
| Quantity | 5 mg |
| Target | Akt (Protein Kinase B) Inhibitors |
| Molecular Formula | C31H27IN4S |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?