missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR7A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13209-1-AP-20UL
This item is not returnable.
View return policy
Description
Aldo-keto reductases, such as AKR7A3, are involved in the detoxification of aldehydes and ketones.Specifications
| AKR7A3 | |
| Polyclonal | |
| Unconjugated | |
| AKR7A3 | |
| AFAR2, AFB1 aldehyde reductase 2, AFB1 AR 2, AKR7A3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22977, 26760 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95154, P38918 | |
| Akr7a3 | |
| AKR7A3 Fusion Protein Ag3991 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction