missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AKR7A3 Monoclonal antibody specifically detects AKR7A3 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | AKR7A3 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 2E11 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | AFAR2, AFB1 aldehyde reductase 2, AFB1-AR 2, aflatoxin B1 aldehyde reductase 2, aflatoxin B1 aldehyde reductase member 3, aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase) |
| Host Species | Mouse |
| Immunogen | Recombinant human AKR7A3 (1-331aa) purified from E. coli |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?