missing translation for 'onlineSavingsMsg'
Learn More

AKR7A3 Antibody (2E11), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18403955 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403955 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403955 Supplier Novus Biologicals Supplier No. NBP197944JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AKR7A3 Monoclonal antibody specifically detects AKR7A3 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR7A3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2E11
Conjugate Janelia Fluor 549
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias AFAR2, AFB1 aldehyde reductase 2, AFB1-AR 2, aflatoxin B1 aldehyde reductase 2, aflatoxin B1 aldehyde reductase member 3, aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase)
Host Species Mouse
Immunogen Recombinant human AKR7A3 (1-331aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 22977
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.