missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR7A3 Antibody (2E11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97944-100UL
This item is not returnable.
View return policy
Description
AKR7A3 Monoclonal specifically detects AKR7A3 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| AKR7A3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| AAH25709 | |
| AKR7A3 | |
| Recombinant human AKR7A3 (1-331aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Anti-human AKR7A3 mAb, clone 2E11, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human AKR7A3 protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 200000000000 | |
| Unconjugated | |
| PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide | |
| AFAR2, AFB1 aldehyde reductase 2, AFB1-AR 2, aflatoxin B1 aldehyde reductase 2, aflatoxin B1 aldehyde reductase member 3, aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 22977 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction