missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKR7A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521565
This item is not returnable.
View return policy
Description
Recommended positive controls: A431. Predicted reactivity: Rat (90%), Pig (91%), Rhesus Monkey (94%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
AKR7A2 is aldo-keto reductases, which are involved in the detoxification of aldehydes and ketones.
Specifications
| AKR7A2 | |
| Polyclonal | |
| Unconjugated | |
| AKR7A2 | |
| 0610025K21Rik; Afar; AFAR1; Afar2; AFB1 aldehyde reductase 1; AFB1-AR 1; AFB1-AR1; aflatoxin aldehyde reductase; aflatoxin B1 aldehyde reductase member 2; aflatoxin beta1 aldehyde reductase; Aiar; AKR7; AKR7A2; Akr7a5; aldo-keto reductase family 7 member A2; aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase); aldo-keto reductase family 7, member A5 (aflatoxin aldehyde reductase); aldoketoreductase 7; epididymis secretory sperm binding protein Li 166mP; HEL-S-166mP; rAFAR2; SSA reductase; succinic semialdehyde reductase; synonyms: AFAR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 110198, 8574 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O43488, Q8CG76 | |
| AKR7A2, Akr7a5 | |
| Recombinant protein encompassing a sequence within the center region of human AKR7A2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction