missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AKR1D1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13403087
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13403087 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13403087 Supplier Invitrogen™ Supplier No. PA528963

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A549. Predicted reactivity: Mouse (82%), Rabbit (89%), Chicken (81%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The enzyme encoded by this gene is responsible for the catalysis of the 5-beta-reduction of bile acid intermediates and steroid hormones carrying a delta(4)-3-one structure. Deficiency of this enzyme may contribute to hepatic dysfunction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR1D1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene AKR1D1
Gene Accession No. P51857
Gene Alias 3o5bred; 3-oxo-5-beta-steroid 4-dehydrogenase; Akr1d1; Aldo-keto reductase family 1 member D1; aldo-keto reductase family 1, member D1; aldo-keto reductase family 1, member D1 (delta 4-3-ketosteroid-5-beta-reductase); CBAS2; delta 4-3-ketosteroid-5-beta-reductase; delta(4)-3-ketosteroid 5-beta-reductase; Delta(4)-3-oxosteroid 5-beta-reductase; SRD5B1; steroid-5-beta-reductase, beta polypeptide 1 (3-oxo-5 beta-steroid delta 4-dehydrogenase beta 1)
Gene Symbols AKR1D1
Host Species Rabbit
Immunogen Full length human AKR1D1 Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6718
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.