missing translation for 'onlineSavingsMsg'
Learn More

AKR1D1 Antibody (1A6), Novus Biologicals™

Product Code. 18319389 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319389 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319389 Supplier Novus Biologicals Supplier No. H00006718M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AKR1D1 Monoclonal antibody specifically detects AKR1D1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR1D1
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 1A6
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005980
Gene Alias 3o5bred, Aldo-keto reductase family 1 member D1,3-oxo-5-beta-steroid 4-dehydrogenase, aldo-keto reductase family 1, member D1 (delta4-3-ketosteroid-5-beta-reductase), CBAS2, Delta(4)-3-oxosteroid 5-beta-reductase, EC 1.3.1.3, SRD5B1beta polypeptide 1 (3-oxo-5 beta-steroid delta4-dehydrogenase beta 1)
Host Species Mouse
Immunogen AKR1D1 (NP_005980, 227 a.a. ~ 326 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NPIWVNVSSPPLLKDALLNSLGKRYNKTAAQIVLRFNIQRGVVVIPKSFNLERIKENFQIFDFSLTEEEMKDIEALNKNVRFVELLMWRDHPEYPFHDEY
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6718
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.