missing translation for 'onlineSavingsMsg'
Learn More

AKR1CL1 Antibody (4G8), Novus Biologicals™

Product Code. 18548330 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18548330 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18548330 Supplier Novus Biologicals Supplier No. H00340811M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AKR1CL1 Monoclonal antibody specifically detects AKR1CL1 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR1CL1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4G8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Alias aldo-keto reductase family 1, member C-like 1, EC 1.1.1.-, FLJ16347
Host Species Mouse
Immunogen AKR1CL1 (NP_001007537.1, 1 a.a. ∽ 129 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MMTDLKQSHSVRLNDGPFMPVLGFGTYAPDHTPKSQAAEATKVAIDVGFRHIDSAYLYQNEEEVGQAIWEKIADGTVKREEIFYTIKLWATFFRAELVHPALERSLKKLGPDYVDLFIIHVPFAMKGSS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340811
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.