missing translation for 'onlineSavingsMsg'
Learn More

AKR1C3-IN-1, MedChemExpress

Product Code. 16495086
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495086 100 mg 100mg
16454118 10 mg 10mg
16450477 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16495086 Supplier Medchem Express Supplier No. HY107379
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AKR1C3-IN-1 is a potent, highly selective inhibitor of AKR1C3, with an IC50 of 13 nM.

AKR1C3-IN-1 is a potent, highly selective inhibitor of AKR1C3, with an IC50 of 13 nM.

Specifications

Chemical Name or Material AKR1C3-IN-1
CAS 327092-81-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H15NO4S
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (315.10 mM)
SMILES O=S(N1CC2=CC=CC=C2CC1)(C3=CC(C(O)=O)=CC=C3)=O
Molecular Weight (g/mol) 317.36
Formula Weight 317.36
Percent Purity 98.74%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.