missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKR1C1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586041
This item is not returnable.
View return policy
Description
AKR1C1 Polyclonal Antibody for Western Blot, IHC (P), Flow
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. These enzymes catalyze the conversion of aldehydes and ketones to their corresponding alcohols by utilizing NADH and/or NADPH as cofactors. The enzymes display overlapping but distinct substrate specificity. This enzyme catalyzes the reaction of progesterone to the inactive form 20-alpha-hydroxy-progesterone. This gene shares high sequence identity with three other gene members and is clustered with those three genes at chromosome 10p15-p14.
Specifications
| AKR1C1 | |
| Polyclonal | |
| Unconjugated | |
| AKR1C1 | |
| 17-beta-HSD 5; 17HSD type 5; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase); 20-ALPHA-HSD; 20-alpha-hydroxysteroid dehydrogenase; 2-ALPHA-HSD; 3alpha-HSD; Akr1c1; Akr1c6; aldo-keto reductase C; aldo-keto reductase family 1 member C1; aldo-keto reductase family 1, member C1; aldo-keto reductase family 1, member C1 (aldehyde reductase); aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase); aldo-keto reductase family 1, member C6; C9; chlordecone reductase homolog HAKRC; DD1; DD1/DD2; DDH; DDH1; dihydrodiol dehydrogenase 1; dihydrodiol dehydrogenase 1/2; dihydrodiol dehydrogenase 1; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase; dihydrodiol dehydrogenase isoform DD1; estradiol 17 beta-dehydrogenase 5; estradiol 17-beta-dehydrogenase (A-specific); H-37; HAKRC; HBAB; hepatic dihydrodiol dehydrogenase; high-affinity hepatic bile acid-binding protein; Hsd17b5; hydroxysteroid 17-beta dehydrogenase 5; indanol dehydrogenase; MBAB; RP11-69D4.1; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; type 5 17beta-hydroxysteroid dehydrogenase; type II 3-alpha-hydroxysteroid dehydrogenase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1645, 83702 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P70694, Q04828 | |
| AKR1C1, Akr1c6 | |
| Recombinant protein within Human AKR1C1 aa 180-323. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction