missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKR1C1/AKR1C2 Recombinant Rabbit Monoclonal Antibody (23GB3175), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553603
This item is not returnable.
View return policy
Description
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. These enzymes catalyze the conversion of aldehydes and ketones to their corresponding alcohols using NADH and/or NADPH as cofactors. The enzymes display overlapping but distinct substrate specificity. This enzyme binds bile acid with high affinity, and shows minimal 3-alpha-hydroxysteroid dehydrogenase activity. And this gene shares high sequence identity with three other gene members and is clustered with those three genes at chromosome 10p15-p14. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| AKR1C1/AKR1C2 | |
| Recombinant Monoclonal | |
| 0.54 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P52895, Q04828 | |
| AKR1C1, Akr1c2 | |
| A synthetic peptide of human AKR1C1/AKR1C2 (50-101AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3175 | |
| Unconjugated | |
| Akr1c2 | |
| 17-alpha-HSD; 17-alpha-hydroxysteroid dehydrogenase; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase); 20-ALPHA-HSD; 20-alpha-hydroxysteroid dehydrogenase; 2-ALPHA-HSD; 3(or 17)-alpha-hydroxysteroid dehydrogenase; 3-alpha-HSD3; 3-alpha-hydroxysteroid dehydrogenase; AKR1C1; Akr1c2; Akr1c21; Akr1c6; AKR1C-pseudo; aldo-keto reductase C; aldo-keto reductase family 1 member C1; Aldo-keto reductase family 1 member C2; aldo-keto reductase family 1 member C21; aldo-keto reductase family 1, member C1; aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase); aldo-keto reductase family 1, member C2; aldo-keto reductase family 1, member C21; aldo-keto reductase family 1, member C6; BABP; C9; chlordecone reductase homolog HAKRC; Chlordecone reductase homolog HAKRD; DD; DD/BABP; DD1; DD1/DD2; DD2; DD-2; DDH; DDH1; DDH2; dihydrodiol dehydrogenase 1; dihydrodiol dehydrogenase 1/2; dihydrodiol dehydrogenase 1; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase; Dihydrodiol dehydrogenase 2; dihydrodiol dehydrogenase 2; bile acid binding protein; 3-alpha hydroxysteroid dehydrogenase, type III; dihydrodiol dehydrogenase isoform DD1; Dihydrodiol dehydrogenase/bile acid-binding protein; H-37; HAKRC; HAKRD; HBAB; hepatic dihydrodiol dehydrogenase; high-affinity hepatic bile acid-binding protein; Hsd17b5; Indanol dehydrogenase; MBAB; MCDR2; pseudo-chlordecone reductase; SRXY8; TDD; testicular 17,20-desmolase deficiency; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; type 5 17beta-hydroxysteroid dehydrogenase; type II 3-alpha-hydroxysteroid dehydrogenase; type II dihydrodiol dehydrogenase; Type III 3-alpha-hydroxysteroid dehydrogenase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1645, 1646 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction