missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1B10 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68327-1-IG-20UL
This item is not returnable.
View return policy
Description
The AKR1B10 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AKR1B10. This antibody recognizes human, pig, rat antigen. The AKR1B10 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| AKR1B10 | |
| Monoclonal | |
| 1000 μg/mL | |
| AKR1B11; AKR1B12; ALDRLn; ARL-1; ARL1; HIS; HSI; MGC14103 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Pig, Rat | |
| 35 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 5A4G5 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 57016 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction