missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKR1A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555680
This item is not returnable.
View return policy
Description
Immunogen sequence: PDEPVLLEEP VVLALAEKYG RSPAQILLRW QVQRKVICIP KSITPSRILQ NIKVFDFTFS PEEMKQLNAL NKNWRYIVPM LTVDGKRVPR DAGHPLYPF Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member, also known as aldehyde reductase, is involved in the reduction of biogenic and xenobiotic aldehydes and is present in virtually every tissue. Multiple alternatively spliced transcript variants of this gene exist, all encoding the same protein.
Specifications
| AKR1A1 | |
| Polyclonal | |
| Unconjugated | |
| AKR1A1 | |
| 2610201A18Rik; 3-DG-reducing enzyme; Akr1a1; AKR1A1 protein; Akr1a4; alcohol dehydrogenase; alcohol dehydrogenase [NADP(+)]; Aldehyde reductase; aldo-keto reductase family 1 member A1; aldo-keto reductase family 1, member A1 (aldehyde reductase); aldo-keto reductase family 1, member A4 (aldehyde reductase); ALDR1; ALR; ARM; DD3; dihydrodiol dehydrogenase 3; epididymis secretory protein Li 6; epididymis secretory sperm binding protein Li 165mP; Glucuronate reductase; Glucuronolactone reductase; HEL-S-165mP; HEL-S-6; MGC12529; MGC1380; OTTHUMP00000009240 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10327, 58810, 78959 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P14550, P51635, Q9JII6 | |
| AKR1A1 | |
| Recombinant protein corresponding to Human AKR1A1. Recombinant protein control fragment (Product #RP-92380). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction