missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AKR1A1 Polyclonal Antibody

Product Code. 13451757
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13451757 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13451757 Supplier Invitrogen™ Supplier No. PA527628

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HepG2. Predicted reactivity: Mouse (92%), Rat (92%), Dog (92%), Pig (94%), Chicken (81%), Rhesus Monkey (100%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member, also known as aldehyde reductase, is involved in the reduction of biogenic and xenobiotic aldehydes and is present in virtually every tissue. Multiple alternatively spliced transcript variants of this gene exist, all encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR1A1
Applications Western Blot
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene AKR1A1
Gene Accession No. P14550
Gene Alias 2610201A18Rik; 3-DG-reducing enzyme; Akr1a1; AKR1A1 protein; Akr1a4; alcohol dehydrogenase; alcohol dehydrogenase [NADP(+)]; Aldehyde reductase; aldo-keto reductase family 1 member A1; aldo-keto reductase family 1, member A1 (aldehyde reductase); aldo-keto reductase family 1, member A4 (aldehyde reductase); ALDR1; ALR; ARM; DD3; dihydrodiol dehydrogenase 3; epididymis secretory protein Li 6; epididymis secretory sperm binding protein Li 165mP; Glucuronate reductase; Glucuronolactone reductase; HEL-S-165mP; HEL-S-6; MGC12529; MGC1380; OTTHUMP00000009240
Gene Symbols AKR1A1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human AKR1A1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10327
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.