missing translation for 'onlineSavingsMsg'
Learn More

AKR1A1 Antibody (OTI6E3), DyLight 594, Novus Biologicals™

Product Code. 18416925 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416925 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416925 Supplier Novus Biologicals Supplier No. NBP271591DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AKR1A1 Monoclonal antibody specifically detects AKR1A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR1A1
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI6E3
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias alcohol dehydrogenase, alcohol dehydrogenase [NADP+], Aldehyde reductase, Aldo-keto reductase family 1 member A1, aldo-keto reductase family 1, member A1 (aldehyde reductase), ALDR1, ALRMGC1380, ARM, DD3, dihydrodiol dehydrogenase 3, EC 1.1.1, EC 1.1.1.2, MGC12529
Host Species Mouse
Immunogen Full length recombinant protein of human AKR1A1 (NP_006057) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10327
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.